Assay Details
Target Gene Details
Entrez Gene ID: | 115350 |
Gene Name: | Fc receptor like 1 |
Gene Aliases: |
CD307a, FCRH1, IFGP1, IRTA5 |
Location: |
Chr.1:157792249-157820150 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCRL1 | NM_001159397.1 | 10 | 1480 | NP_001152869.1 |
| NM_001159398.1 | 11 | 1604 | NP_001152870.1 | |
| NM_052938.4 | 11 | 1607 | NP_443170.1 | |
| XM_005244866.2 | 11 | 1568 | XP_005244923.1 | |
| XM_005244867.2 | 10 | 1461 | XP_005244924.1 | |
| XM_005244869.3 | 9 | 1303 | XP_005244926.1 | |
| XM_011509125.1 | 12 | 1576 | XP_011507427.1 | |
| XM_011509126.1 | 12 | 1573 | XP_011507428.1 | |
| XM_011509127.2 | 10 | 1570 | XP_011507429.1 | |
| XM_011509128.1 | 11 | 1580 | XP_011507430.1 | |
| XM_011509129.1 | 11 | 1577 | XP_011507431.1 | |
| XM_011509130.1 | 11 | 1565 | XP_011507432.1 | |
| XM_011509133.2 | 11 | 1432 | XP_011507435.1 | |
| XM_011509134.2 | 10 | 1407 | XP_011507436.1 | |
| XM_011509135.2 | 10 | 1324 | XP_011507437.1 | |
| XM_011509137.2 | 8 | 952 | XP_011507439.1 | |
| XM_017000227.1 | 10 | 3574 | XP_016855716.1 | |
| AF329488.1 | 11 | 1514 | AAL23898.1 | |
| AF459634.1 | 11 | 1560 | AAL60250.1 | |
| AK096690.1 | 10 | 1419 | BAC04842.1 | |
| AL833674.1 | 7 | 651 | ||
| AL833970.1 | 7 | 1595 | CAD38815.1 | |
| AY043464.1 | 11 | 1562 | AAK91777.1 | |
| BC033690.1 | 11 | 1558 | AAH33690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587657 | Chr.1:157793395 - 157801763 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map