Assay Details
Target Gene Details
Entrez Gene ID: | 5188 |
Gene Name: | glutamyl-tRNA amidotransferase subunit B |
Gene Aliases: |
HSPC199, PET112, PET112L |
Location: |
Chr.4:151669773-151761023 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GATB | NM_004564.2 | 13 | 1662 | NP_004555.1 |
| XM_005263063.3 | 12 | 1489 | XP_005263120.1 | |
| AB019410.2 | 13 | 1606 | ||
| AF026851.1 | 13 | 1633 | AAD08640.1 | |
| AF151033.1 | 13 | 1628 | AAF36119.1 | |
| AK124986.1 | 2 | 3355 | ||
| AK222887.1 | 13 | 1624 | BAD96607.1 | |
| AK312957.1 | 13 | 1624 | ||
| BC130348.1 | 13 | 1672 | ||
| BC136547.1 | 13 | 1672 | ||
| BX537555.1 | 11 | 6174 | CAH56162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4551 | Chr.4:151661465 - 151706215 on Build GRCh38 | Deletion |
|
| nsv1028814 | Chr.4:150874356 - 152285445 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map