Assay Details
Target Gene Details
Entrez Gene ID: | 6173 |
Gene Name: | ribosomal protein L36a |
Gene Aliases: |
L36A, L44L, MIG6, RPL44 |
Location: |
Chr.X:101390890-101396154 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL36A | NM_021029.5 | 5 | 586 | NP_066357.2 |
| AW150895.1 | 1 | 296 | ||
| BG504049.1 | 5 | 465 |
Target Gene Details
Entrez Gene ID: | 100529097 |
Gene Name: | RPL36A-HNRNPH2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.X:101390890-101414140 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL36A-HNRNPH2 | NM_001199973.1 | NP_001186902.1 | ||
| NM_001199974.1 | NP_001186903.1 | |||
| DA530036.1 | ||||
| DA998453.1 |
More Information
Set Membership: |
|

Genomic Map