Assay Details
Target Gene Details
Entrez Gene ID: | 84129 |
Gene Name: | acyl-CoA dehydrogenase family member 11 |
Gene Aliases: |
ACAD-11 |
Location: |
Chr.3:132558138-132660131 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAD11 | NM_032169.4 | NP_115545.3 | ||
| NR_132426.1 | ||||
| NR_132427.1 | ||||
| NR_132428.1 | ||||
| AI091999.1 | ||||
| AL833721.1 | CAH56228.1 | |||
| BC019607.1 | AAH19607.2 | |||
| CB240680.1 |
Target Gene Details
Entrez Gene ID: | 100532724 |
Gene Name: | NPHP3-ACAD11 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.3:132558138-132722459 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 45 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPHP3-ACAD11 | NR_037804.1 | |||
| AB082531.1 | BAC02709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591797 | Chr.3:132554869 - 132565301 on Build GRCh38 | Loss |
|
| esv3597827 | Chr.3:132554241 - 132561393 on Build GRCh38 | Loss |
|
| nsv965409 | Chr.3:132553912 - 132561059 on Build GRCh38 | Deletion |
|
| nsv591796 | Chr.3:132552294 - 132559581 on Build GRCh38 | Loss |
|
| esv3569006 | Chr.3:132550754 - 132562086 on Build GRCh38 | Loss |
|
| nsv829728 | Chr.3:132359717 - 132563778 on Build GRCh38 | Gain |
|
| esv2675190 | Chr.3:132554212 - 132561501 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map