Assay Details
Target Gene Details
Entrez Gene ID: | 9318 |
Gene Name: | COP9 signalosome subunit 2 |
Gene Aliases: |
ALIEN, CSN2, SGN2, TRIP15 |
Location: |
Chr.15:49125274-49155657 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPS2 | NM_001143887.1 | NP_001137359.1 | ||
| NM_004236.3 | NP_004227.1 | |||
| AB209799.1 | BAD93036.1 | |||
| AF084260.1 | AAC34122.1 | |||
| AF100762.1 | AAD43026.1 | |||
| AF120268.1 | AAD30269.1 | |||
| AF212227.1 | AAK26250.1 | |||
| AK222590.1 | BAD96310.1 | |||
| AK295598.1 | ||||
| AK312643.1 | ||||
| BC012629.1 | AAH12629.1 | |||
| CR542063.1 | CAG46860.1 | |||
| DB059510.1 | ||||
| L40388.1 | AAC41734.1 |
More Information
Set Membership: |
|

Genomic Map