Assay Details
Target Gene Details
Entrez Gene ID: | 10501 |
Gene Name: | semaphorin 6B |
Gene Aliases: |
SEM-SEMA-Y, SEMA-VIB, SEMAN, semaZ |
Location: |
Chr.19:4542588-4581484 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA6B | NM_032108.3 | 17 | 3908 | NP_115484.2 |
| XM_011527639.2 | 17 | 3975 | XP_011525941.1 | |
| XM_011527640.2 | 17 | 3718 | XP_011525942.1 | |
| AB209278.1 | BAD92515.1 | |||
| AI825930.1 | 1 | 54 | ||
| AK300037.1 | ||||
| AY358939.1 | AAQ89298.1 | |||
| BC142617.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828405 | Chr.19:4540168 - 4546225 on Build GRCh38 | Gain |
|
| dgv614e199 | Chr.19:4541633 - 4547580 on Build GRCh38 | Deletion |
|
| nsv953950 | Chr.19:4540589 - 4723488 on Build GRCh38 | Deletion |
|
| nsv578444 | Chr.19:4502189 - 4559634 on Build GRCh38 | Gain |
|
| nsv428361 | Chr.19:4510042 - 4689189 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map