Assay Details
Target Gene Details
Entrez Gene ID: | 55739 | 
Gene Name: | NAD(P)HX dehydratase | 
Gene Aliases: | 
	
		CARKD, LP3298 | 
Location: | 
		
			Chr.13:110615460-110639996 on Build GRCh38 | 
		Assay Gene Location: | Within Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| NAXD | NM_001242881.1 | 2 | 266 | NP_001229810.1 | 
| NM_001242882.1 | 2 | 263 | NP_001229811.1 | |
| NM_001242883.1 | NP_001229812.1 | |||
| NM_018210.3 | 2 | 266 | NP_060680.2 | |
| NR_040103.1 | 2 | 263 | ||
| NR_040104.1 | 2 | 263 | ||
| AK001631.1 | 2 | 189 | BAA91797.1 | |
| AK024260.1 | 2 | 132 | BAB14863.1 | |
| AK057767.1 | 2 | 146 | ||
| AK093771.1 | 2 | 189 | ||
| AK225129.1 | 2 | 188 | ||
| AK296227.1 | 2 | 140 | ||
| AK296758.1 | 2 | 147 | ||
| AK298917.1 | ||||
| AK302117.1 | ||||
| BC041028.1 | 2 | 197 | AAH41028.1 | |
| BQ955656.1 | 2 | 162 | ||
| DA175011.1 | 2 | 259 | ||
| DA435066.1 | 2 | 269 | ||
| DA815628.1 | 2 | 263 | 
Target Copy Number Variation Details
	    DGV Version: |  
	    Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation  |  
		            Location | CNV Subtype  |  
		            Genes | 
|---|---|---|---|
| nsv1038089 | Chr.13:110577170 - 110622476 on Build GRCh38 | Loss | 
							
								 | 
					
| nsv1044234 | Chr.13:110490706 - 110718233 on Build GRCh38 | Gain | 
							
								 | 
					
| nsv563124 | Chr.13:110608920 - 110648596 on Build GRCh38 | Loss | 
							
								 | 
					
| nsv1046423 | Chr.13:110283757 - 110634260 on Build GRCh38 | Gain | 
							
								 | 
					
| nsv563123 | Chr.13:110584913 - 110639697 on Build GRCh38 | Loss | 
							
								 | 
					
| nsv832720 | Chr.13:110447888 - 110643646 on Build GRCh38 | Loss | 
							
								 | 
					
| nsv1044340 | Chr.13:110581726 - 111040647 on Build GRCh38 | Gain | 
							
								 | 
					
More Information
Set Membership: | 
		   
					
						
							 | 
				

Genomic Map