Assay Details
Target Gene Details
Entrez Gene ID: | 6283 |
Gene Name: | S100 calcium binding protein A12 |
Gene Aliases: |
CAAF1, CAGC, CGRP, ENRAGE, MRP-6, MRP6, p6 |
Location: |
Chr.1:153373708-153375599 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A12 | NM_005621.1 | NP_005612.1 | ||
| BC070294.1 | AAH70294.1 | |||
| D83664.1 | BAA12036.1 | |||
| X97859.1 | CAA66453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002705 | Chr.1:153335376 - 153553113 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map