Assay Details
Target Gene Details
Entrez Gene ID: | 65258 |
Gene Name: | metallophosphoesterase 1 |
Gene Aliases: |
PGAP5 |
Location: |
Chr.18:11883471-11908797 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057995 | Chr.18:10626172 - 11923368 on Build GRCh38 | Gain |
|
| nsv833590 | Chr.18:11757330 - 11924540 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map