Assay Details
Target Gene Details
Entrez Gene ID: | 130872 |
Gene Name: | AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast) |
Gene Aliases: |
Hch1 |
Location: |
Chr.2:61177418-61191210 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHSA2 | NM_001321300.1 | 7 | 1669 | NP_001308229.1 |
| NM_001321301.1 | 7 | 2381 | NP_001308230.1 | |
| NM_001321302.1 | 8 | 1089 | NP_001308231.1 | |
| NM_001321303.1 | 7 | 2367 | NP_001308232.1 | |
| NM_001321304.1 | 5 | 3092 | NP_001308233.1 | |
| NM_001321305.1 | NP_001308234.1 | |||
| NM_152392.4 | 6 | 2265 | NP_689605.1 | |
| XM_011532541.2 | 8 | 1325 | XP_011530843.1 | |
| XM_011532543.2 | 9 | 1609 | XP_011530845.1 | |
| XM_011532545.2 | XP_011530847.1 | |||
| XM_017003377.1 | XP_016858866.1 | |||
| XM_017003378.1 | 9 | 1284 | XP_016858867.1 | |
| AF542548.1 | 5 | 466 | AAQ09531.1 | |
| AK091998.1 | 1 | 505 | ||
| AK092505.1 | ||||
| AK092817.1 | 6 | 1669 | ||
| AL833813.1 | 6 | 2000 | CAD38676.1 | |
| BC113671.1 | ||||
| BC117278.1 | ||||
| BX505714.1 | 2 | 226 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760616 | Chr.2:61136348 - 61200572 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map