Assay Details
Target Gene Details
Entrez Gene ID: | 9331 |
Gene Name: | beta-1,4-galactosyltransferase 6 |
Gene Aliases: |
B4Gal-T6, beta4Gal-T6 |
Location: |
Chr.18:31622246-31724670 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT6 | NM_004775.3 | 9 | 3784 | NP_004766.2 |
| XM_005258387.4 | 9 | 3549 | XP_005258444.1 | |
| XM_006722578.3 | 8 | 4638 | XP_006722641.1 | |
| XM_006722579.3 | 10 | 3497 | XP_006722642.1 | |
| XM_017026088.1 | 8 | 4635 | XP_016881577.1 | |
| XM_017026089.1 | 9 | 3955 | XP_016881578.1 | |
| XM_017026090.1 | 7 | 5398 | XP_016881579.1 | |
| XM_017026091.1 | 7 | 3252 | XP_016881580.1 | |
| XM_017026092.1 | 8 | 3830 | XP_016881581.1 | |
| AB209194.1 | 5 | 3117 | BAD92431.1 | |
| AF097159.1 | 9 | 3784 | AAD41695.1 |
More Information
Set Membership: |
|

Genomic Map