Assay Details
Target Gene Details
Entrez Gene ID: | 57595 |
Gene Name: | PDZ domain containing 4 |
Gene Aliases: |
LNX5, LU1, PDZK4, PDZRN4L |
Location: |
Chr.X:153802166-153830567 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDZD4 | NM_001303512.1 | NP_001290441.1 | ||
| NM_001303513.1 | NP_001290442.1 | |||
| NM_001303514.1 | NP_001290443.1 | |||
| NM_001303515.1 | NP_001290444.1 | |||
| NM_001303516.1 | NP_001290445.1 | |||
| NM_032512.3 | NP_115901.2 | |||
| AB103478.1 | BAD16631.1 | |||
| AK091444.1 | BAC03666.1 | |||
| AK123822.1 | ||||
| AK127016.1 | ||||
| AK309212.1 | ||||
| BC117277.1 | ||||
| BC143459.1 | ||||
| BC143545.1 | ||||
| DA097500.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|

Genomic Map