Assay Details
Target Gene Details
Entrez Gene ID: | 56853 |
Gene Name: | CUGBP, Elav-like family member 4 |
Gene Aliases: |
BRUNOL4, CELF-4 |
Location: |
Chr.18:37243040-37566052 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELF4 | NM_001025087.1 | 13 | 3034 | NP_001020258.1 |
| NM_001025088.1 | 13 | 3031 | NP_001020259.1 | |
| NM_001025089.1 | 12 | 2923 | NP_001020260.1 | |
| NM_020180.3 | 13 | 3037 | NP_064565.1 | |
| XM_017025866.1 | 13 | 2469 | XP_016881355.1 | |
| BM718619.1 | 1 | 305 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833621 | Chr.18:37127725 - 37302713 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map