Assay Details
Target Gene Details
Entrez Gene ID: | 10332 |
Gene Name: | C-type lectin domain family 4 member M |
Gene Aliases: |
CD209L, CD299, DC-SIGN2, DC-SIGNR, DCSIGNR, HP10347, L-SIGN, LSIGN |
Location: |
Chr.19:7763052-7770559 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC4M | NM_001144904.1 | NP_001138376.1 | ||
| NM_001144905.1 | NP_001138377.1 | |||
| NM_001144906.1 | NP_001138378.1 | |||
| NM_001144907.1 | NP_001138379.1 | |||
| NM_001144908.1 | NP_001138380.1 | |||
| NM_001144909.1 | NP_001138381.1 | |||
| NM_001144910.1 | NP_001138382.1 | |||
| NM_001144911.1 | NP_001138383.1 | |||
| NM_014257.4 | NP_055072.3 | |||
| NR_026707.1 | 4 | 1103 | ||
| NR_026708.1 | 4 | 1103 | ||
| NR_026709.1 | 3 | 1040 | ||
| XM_006722611.2 | XP_006722674.1 | |||
| XM_006722612.3 | XP_006722675.1 | |||
| XM_006722613.3 | XP_006722676.1 | |||
| XM_006722614.3 | XP_006722677.1 | |||
| XM_006722615.2 | XP_006722678.1 | |||
| AB015629.1 | BAA76496.1 | |||
| AF245219.2 | AAG13848.2 | |||
| AF290887.1 | AAK20998.1 | |||
| AK023750.1 | 2 | 824 | BAB14667.1 | |
| AK292278.1 | ||||
| AK304499.1 | ||||
| AK314704.1 | ||||
| AY042234.1 | AAK91859.1 | |||
| AY042235.1 | 4 | 985 | AAK91860.1 | |
| AY042236.1 | 4 | 985 | AAK91861.1 | |
| AY042237.1 | AAK91862.1 | |||
| AY042238.1 | AAK91863.1 | |||
| AY042239.1 | 3 | 922 | AAK91864.1 | |
| AY042240.1 | AAK91865.1 | |||
| AY343913.1 | AAR04559.1 | |||
| BC038851.1 | AAH38851.1 | |||
| BC110614.2 | AAI10615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060887 | Chr.19:7089546 - 7861087 on Build GRCh38 | Gain |
|
| nsv833732 | Chr.19:7672656 - 7871329 on Build GRCh38 | Gain |
|
| esv2751809 | Chr.19:6896369 - 7816144 on Build GRCh38 | Gain |
|
| nsv1061452 | Chr.19:7657750 - 7785131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map