Assay Details
Target Gene Details
| Entrez Gene ID: | 11126 | 
| Gene Name: | CD160 molecule | 
| Gene Aliases: | BY55, NK1, NK28 | 
| Location: | Chr.1:145719312-145739288 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CD160 | NM_007053.3 | 6 | 870 | NP_008984.1 | 
| NR_103845.1 | 4 | 398 | ||
| AF060981.1 | 6 | 793 | AAC72302.1 | |
| AK128370.1 | 3 | 2760 | ||
| BC014465.1 | 6 | 796 | AAH14465.1 | |
| CD638675.1 | 4 | 401 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3587421 | Chr.1:145649153 - 145751059 on Build GRCh38 | Loss |  RNF115  CD160  GPR89A  PDZK1 | 
| dgv331n100 | Chr.1:145642988 - 145827134 on Build GRCh38 | Gain |  RNF115  CD160  GPR89A  PDZK1  POLR3C | 
| esv3572342 | Chr.1:145601946 - 145809476 on Build GRCh38 | Gain |  RNF115  CD160  NBPF25P  GPR89A  PDZK1 | 
| nsv469752 | Chr.1:145639438 - 145790042 on Build GRCh38 | Loss |  RNF115  CD160  GPR89A  PDZK1 | 
| nsv831392 | Chr.1:145632374 - 145813165 on Build GRCh38 | Gain |  RNF115  CD160  GPR89A  PDZK1 | 
| esv3894089 | Chr.1:145672100 - 145824903 on Build GRCh38 | Loss |  RNF115  CD160  PDZK1  POLR3C | 
| nsv547670 | Chr.1:145689573 - 145778504 on Build GRCh38 | Loss |  RNF115  CD160  PDZK1 | 
| dgv532n54 | Chr.1:145624561 - 145813526 on Build GRCh38 | Gain |  RNF115  CD160  GPR89A  PDZK1 | 
| dgv33n27 | Chr.1:145687602 - 145824903 on Build GRCh38 | Loss |  RNF115  CD160  PDZK1  POLR3C | 
| dgv95e212 | Chr.1:145631218 - 145812561 on Build GRCh38 | Gain |  RNF115  CD160  GPR89A  PDZK1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top