Assay Details
Target Gene Details
Entrez Gene ID: | 2060 |
Gene Name: | epidermal growth factor receptor pathway substrate 15 |
Gene Aliases: |
AF-1P, AF1P, MLLT5 |
Location: |
Chr.1:51354263-51519354 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPS15 | NM_001159969.1 | 13 | 2216 | NP_001153441.1 |
| NM_001981.2 | 25 | 3085 | NP_001972.1 | |
| XM_005270618.2 | 24 | 3026 | XP_005270675.1 | |
| XM_017000615.1 | 25 | 3227 | XP_016856104.1 | |
| XM_017000616.1 | 24 | 3023 | XP_016856105.1 | |
| XM_017000617.1 | 24 | 2912 | XP_016856106.1 | |
| XM_017000618.1 | 23 | 2822 | XP_016856107.1 | |
| XM_017000619.1 | 26 | 3109 | XP_016856108.1 | |
| XM_017000620.1 | 25 | 3276 | XP_016856109.1 | |
| AF052132.1 | 2 | 605 | ||
| AK090936.1 | 13 | 1942 | ||
| AK294297.1 | 14 | 2065 | ||
| BC054006.1 | 24 | 2601 | AAH54006.1 | |
| BX647676.1 | 13 | 2197 | ||
| U07707.1 | 25 | 3014 | AAA52101.1 | |
| Z29064.1 | 25 | 3085 | CAA82305.1 |
More Information
Set Membership: |
|

Genomic Map