Assay Details
Target Gene Details
Entrez Gene ID: | 10576 |
Gene Name: | chaperonin containing TCP1 subunit 2 |
Gene Aliases: |
99D8.1, CCT-beta, CCTB, HEL-S-100n, PRO1633, TCP-1-beta |
Location: |
Chr.12:69585428-69601577 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCT2 | NM_001198842.1 | 7 | 871 | NP_001185771.1 |
| NM_006431.2 | 7 | 640 | NP_006422.1 | |
| AB451223.1 | 6 | 546 | ||
| AB451346.1 | 6 | 546 | ||
| AF026166.1 | 7 | 603 | AAC98906.1 | |
| AF026293.1 | 7 | 579 | AAC96012.1 | |
| AK290767.1 | 7 | 635 | ||
| AK294307.1 | 7 | 885 | ||
| AK297685.1 | 5 | 427 | ||
| AK302157.1 | 7 | 680 | ||
| AK316397.1 | 7 | 810 | ||
| AK316408.1 | 7 | 871 | ||
| BC113514.1 | 7 | 618 | ||
| BC113516.1 | 7 | 618 | ||
| BT019966.1 | 6 | 546 | AAV38769.1 | |
| CR457071.1 | 6 | 546 | CAG33352.1 | |
| FJ224351.1 | 7 | 579 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1523n100 | Chr.12:69523935 - 69598862 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map