Assay Details
Target Gene Details
Entrez Gene ID: | 127495 |
Gene Name: | leucine rich repeat containing 39 |
Gene Aliases: |
- |
Location: |
Chr.1:100148448-100178273 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC39 | NM_001256385.1 | 11 | 1407 | NP_001243314.1 |
| NM_001256386.1 | 10 | 1337 | NP_001243315.1 | |
| NM_001256387.1 | 9 | 1280 | NP_001243316.1 | |
| NM_144620.3 | 10 | 1320 | NP_653221.1 | |
| AK096892.1 | ||||
| AL833417.1 | 11 | 1355 | ||
| AY358131.1 | AAQ88498.1 | |||
| BC009613.1 | 10 | 1262 | AAH09613.1 | |
| DB449006.1 | 1 | 280 |
Target Gene Details
Entrez Gene ID: | 54482 |
Gene Name: | tRNA methyltransferase 13 homolog |
Gene Aliases: |
CCDC76 |
Location: |
Chr.1:100133080-100150498 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT13 | NM_019083.2 | 11 | 1632 | NP_061956.2 |
| NR_135078.1 | 11 | 1391 | ||
| XM_005270945.3 | 11 | 1660 | XP_005271002.1 | |
| XM_005270946.2 | 10 | 1552 | XP_005271003.1 | |
| XM_017001504.1 | 10 | 3284 | XP_016856993.1 | |
| XM_017001505.1 | 10 | 3273 | XP_016856994.1 | |
| AK002081.1 | 11 | 1619 | BAA92074.1 | |
| BC075811.1 | 11 | 1629 | AAH75811.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2121 | Chr.1:100115063 - 100149647 on Build GRCh38 | Insertion |
|
| nsv830804 | Chr.1:100036777 - 100210481 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map