Assay Details
Target Gene Details
Entrez Gene ID: | 84945 |
Gene Name: | abhydrolase domain containing 13 |
Gene Aliases: |
BEM46L1, C13orf6, bA153I24.2 |
Location: |
Chr.13:108218415-108235440 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD13 | NM_032859.2 | 2 | 2492 | NP_116248.2 |
| XM_011521128.2 | 2 | 2450 | XP_011519430.1 | |
| AK027812.1 | 1 | 1949 | BAB55387.1 | |
| BX647428.1 | 1 | 2072 |
More Information
Set Membership: |
|

Genomic Map