Assay Details
Target Gene Details
Entrez Gene ID: | 317649 |
Gene Name: | eukaryotic translation initiation factor 4E family member 3 |
Gene Aliases: |
eIF-4E3, eIF4E-3 |
Location: |
Chr.3:71679289-71754831 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4E3 | NM_001134649.2 | 8 | 2775 | NP_001128121.1 |
| NM_001134650.1 | 8 | 2651 | NP_001128122.1 | |
| NM_001134651.1 | 7 | 2642 | NP_001128123.1 | |
| NM_001282886.1 | 8 | 2768 | NP_001269815.1 | |
| NM_173359.4 | 8 | 2736 | NP_775495.1 | |
| XM_011533651.2 | 5 | 2495 | XP_011531953.1 | |
| XM_017006278.1 | 9 | 2782 | XP_016861767.1 | |
| XM_017006279.1 | 9 | 2769 | XP_016861768.1 | |
| XM_017006280.1 | 9 | 2916 | XP_016861769.1 | |
| XM_017006281.1 | 7 | 2644 | XP_016861770.1 | |
| XM_017006282.1 | 9 | 2724 | XP_016861771.1 | |
| AK126999.1 | 7 | 2524 | ||
| AL161983.1 | 1 | 1086 | ||
| BC068443.1 | AAH68443.1 | |||
| BX648727.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999942 | Chr.3:71131662 - 71736899 on Build GRCh38 | Gain |
|
| nsv834729 | Chr.3:71617352 - 71694991 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map