Assay Details
Target Gene Details
Entrez Gene ID: | 6574 |
Gene Name: | solute carrier family 20 member 1 |
Gene Aliases: |
GLVR1, Glvr-1, PIT1, PiT-1 |
Location: |
Chr.2:112645857-112663825 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC20A1 | NM_005415.4 | 11 | 3124 | NP_005406.3 |
| XM_017004768.1 | 11 | 2873 | XP_016860257.1 | |
| XM_017004769.1 | 12 | 5351 | XP_016860258.1 | |
| XM_017004770.1 | 12 | 3154 | XP_016860259.1 | |
| XM_017004771.1 | 13 | 3190 | XP_016860260.1 | |
| AK123420.1 | 5 | 2161 | ||
| BC019944.1 | 11 | 3031 | AAH19944.1 | |
| BC075818.1 | 5 | 1633 | AAH75818.1 | |
| BX647555.1 | 3 | 2953 | ||
| L20859.1 | 11 | 2955 | AAA52572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458997 | Chr.2:112556217 - 112673515 on Build GRCh38 | Gain |
|
| nsv470480 | Chr.2:112616373 - 112672175 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map