Assay Details
Target Gene Details
Entrez Gene ID: | 166647 |
Gene Name: | adhesion G protein-coupled receptor A3 |
Gene Aliases: |
GPR125, PGR21, TEM5L |
Location: |
Chr.4:22387374-22516058 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRA3 | NM_145290.3 | NP_660333.2 | ||
| XM_011513811.2 | XP_011512113.1 | |||
| XM_017007834.1 | XP_016863323.1 | |||
| AI198439.1 | ||||
| AK095866.1 | ||||
| AK299168.1 | ||||
| AL832257.1 | ||||
| BX648287.1 |
More Information
Set Membership: |
|

Genomic Map