Assay Details
Target Gene Details
Entrez Gene ID: | 83694 |
Gene Name: | ribosomal protein S6 kinase like 1 |
Gene Aliases: |
RSKL2 |
Location: |
Chr.14:74903954-74923396 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KL1 | NM_031464.4 | NP_113652.2 | ||
| XM_011537206.2 | XP_011535508.1 | |||
| XM_011537207.2 | XP_011535509.1 | |||
| XM_011537211.2 | XP_011535513.1 | |||
| XM_011537213.2 | XP_011535515.1 | |||
| XM_017021680.1 | XP_016877169.1 | |||
| XM_017021681.1 | XP_016877170.1 | |||
| XM_017021682.1 | XP_016877171.1 | |||
| XM_017021683.1 | XP_016877172.1 | |||
| XM_017021684.1 | XP_016877173.1 | |||
| XM_017021685.1 | XP_016877174.1 | |||
| XM_017021686.1 | XP_016877175.1 | |||
| XM_017021687.1 | XP_016877176.1 | |||
| XM_017021688.1 | XP_016877177.1 | |||
| XM_017021689.1 | XP_016877178.1 | |||
| XM_017021690.1 | XP_016877179.1 | |||
| XM_017021691.1 | XP_016877180.1 | |||
| XM_017021692.1 | XP_016877181.1 | |||
| AK131532.1 | 1 | 539 | ||
| AK226137.1 | ||||
| AK299846.1 | ||||
| BC004540.2 | AAH04540.1 |
More Information
Set Membership: |
|

Genomic Map