Assay Details
Target Gene Details
Entrez Gene ID: | 60436 |
Gene Name: | TGFB induced factor homeobox 2 |
Gene Aliases: |
- |
Location: |
Chr.20:36573473-36593952 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGIF2 | NM_001199513.1 | 3 | 721 | NP_001186442.1 |
| NM_001199514.1 | 3 | 849 | NP_001186443.1 | |
| NM_001199515.1 | 3 | 751 | NP_001186444.1 | |
| NM_021809.6 | 3 | 808 | NP_068581.1 | |
| AB042646.1 | 3 | 817 | BAB16424.1 | |
| AK074802.1 | 3 | 788 | BAC11216.1 | |
| AK074829.1 | 1 | 303 | ||
| AK313919.1 | 3 | 808 | ||
| BC006549.2 | 3 | 795 | AAH06549.1 | |
| BC012816.2 | 3 | 795 | AAH12816.1 |
Target Gene Details
Entrez Gene ID: | 100527943 |
Gene Name: | TGIF2-C20orf24 readthrough |
Gene Aliases: |
- |
Location: |
Chr.20:36574554-36612557 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGIF2-C20orf24 | NM_001199535.1 | NP_001186464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss |
|
| nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map