Assay Details
Target Gene Details
Entrez Gene ID: | 9935 |
Gene Name: | MAF bZIP transcription factor B |
Gene Aliases: |
KRML, MCTO |
Location: |
Chr.20:40685848-40689240 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAFB | NM_005461.4 | 1 | 2195 | NP_005452.2 |
| AF086178.1 | 1 | 223 | ||
| AF134157.1 | 1 | 1878 | AAD30106.1 | |
| AK027324.1 | 2 | 1734 | ||
| BC028098.1 | 1 | 1826 | AAH28098.1 | |
| BC036689.1 | 1 | 2196 | AAH36689.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064246 | Chr.20:40261343 - 42254320 on Build GRCh38 | Loss |
|
| nsv833978 | Chr.20:40684655 - 40881584 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map