Assay Details
Target Gene Details
Entrez Gene ID: | 9736 |
Gene Name: | ubiquitin specific peptidase 34 |
Gene Aliases: |
- |
Location: |
Chr.2:61187455-61470714 on Build GRCh38 |
Assay Gene Location: | Within Exon 69 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP34 | NM_014709.3 | 69 | 8807 | NP_055524.3 |
| AB011142.2 | 70 | 8862 | BAA25496.2 | |
| AB018272.1 | 12 | 1736 | BAA34449.1 | |
| AJ586138.1 | 67 | 8332 | CAE51938.1 | |
| AK001193.1 | 9 | 1398 | ||
| AK023171.1 | 1 | 84 | ||
| AK023845.1 | 19 | 2212 | ||
| AK125898.1 | 28 | 3326 | ||
| AL050092.1 | 4 | 903 | CAB43264.1 | |
| BC022783.1 | 6 | 1106 | AAH22783.1 |
More Information
Set Membership: |
|

Genomic Map