Assay Details
Target Gene Details
Entrez Gene ID: | 239 |
Gene Name: | arachidonate 12-lipoxygenase, 12S type |
Gene Aliases: |
12-LOX, 12S-LOX, LOG12 |
Location: |
Chr.17:6993791-7010736 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALOX12 | NM_000697.2 | NP_000688.2 | ||
| XM_011523780.2 | XP_011522082.1 | |||
| BC069557.1 | AAH69557.1 | |||
| M35418.1 | AAA60056.1 | |||
| M58704.1 | AAA59523.1 | |||
| M62982.1 | AAA51533.1 |
Target Gene Details
Entrez Gene ID: | 100506713 |
Gene Name: | ALOX12 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:6985123-7012334 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALOX12-AS1 | NR_040089.1 | |||
| BI559919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| esv1008363 | Chr.17:7005907 - 7006407 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map