Assay Details
Target Gene Details
Entrez Gene ID: | 84269 |
Gene Name: | coiled-coil-helix-coiled-coil-helix domain containing 5 |
Gene Aliases: |
C2orf9, MIC14 |
Location: |
Chr.2:112583606-112589040 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHCHD5 | NM_001304353.1 | 2 | 140 | NP_001291282.1 |
| NM_001304354.1 | 2 | 137 | NP_001291283.1 | |
| NM_032309.3 | 2 | 303 | NP_115685.1 | |
| XM_005263824.3 | 2 | 1134 | XP_005263881.1 | |
| AK096990.1 | 2 | 145 | BAC04922.1 | |
| AK312219.1 | 2 | 279 | ||
| BC004498.2 | 2 | 281 | AAH04498.1 | |
| BM563093.1 | 2 | 137 | ||
| BX115079.1 | 2 | 139 | ||
| HY011716.1 | 2 | 298 | ||
| N39730.1 | 2 | 140 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458997 | Chr.2:112556217 - 112673515 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map