Assay Details
Target Gene Details
Entrez Gene ID: | 8500 |
Gene Name: | PTPRF interacting protein alpha 1 |
Gene Aliases: |
LIP.1, LIP1, LIPRIN |
Location: |
Chr.11:70270687-70384501 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Set Membership: |
|

Genomic Map