Assay Details
Target Gene Details
Entrez Gene ID: | 836 |
Gene Name: | caspase 3 |
Gene Aliases: |
CPP32, CPP32B, SCA-1 |
Location: |
Chr.4:184627696-184649475 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASP3 | NM_004346.3 | 8 | 1495 | NP_004337.2 |
| NM_032991.2 | 7 | 1328 | NP_116786.1 | |
| XM_011532301.1 | 8 | 1429 | XP_011530603.1 | |
| AK291337.1 | 8 | 1466 | ||
| BC016926.2 | 8 | 1447 | AAH16926.1 | |
| BX647609.1 | 7 | 1447 | CAI46084.1 | |
| U13737.1 | 8 | 1456 | AAA65015.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830168 | Chr.4:184626297 - 184801578 on Build GRCh38 | Loss |
|
| nsv1026074 | Chr.4:184384963 - 184780663 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map