Assay Details
Target Gene Details
Entrez Gene ID: | 10097 |
Gene Name: | ARP2 actin related protein 2 homolog |
Gene Aliases: |
ARP2 |
Location: |
Chr.2:65227695-65271253 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR2 | NM_001005386.2 | 1 | 135 | NP_001005386.1 |
| NM_005722.3 | 1 | 135 | NP_005713.1 | |
| AF006082.1 | AAB64187.1 | |||
| AK295157.1 | 1 | 85 | ||
| AK301634.1 | ||||
| AK315205.1 | ||||
| BC014546.1 | AAH14546.1 | |||
| BI670123.1 | 1 | 72 | ||
| BX491447.1 | 1 | 39 | ||
| BX649080.1 | ||||
| DC404298.1 | 1 | 135 |
More Information
Set Membership: |
|

Genomic Map