Assay Details
Target Gene Details
Entrez Gene ID: | 6730 |
Gene Name: | signal recognition particle 68 |
Gene Aliases: |
- |
Location: |
Chr.17:76038775-76072526 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRP68 | NM_001260502.1 | 15 | 1976 | NP_001247431.1 |
| NM_001260503.1 | 9 | 1285 | NP_001247432.1 | |
| NM_014230.3 | 16 | 2090 | NP_055045.2 | |
| NR_048541.1 | 15 | 1986 | ||
| AK002002.1 | 11 | 1545 | ||
| AK027320.1 | BAB55040.1 | |||
| AK074698.1 | 16 | 2066 | BAC11145.1 | |
| AK092935.1 | 2 | 1609 | ||
| AK097962.1 | 9 | 1285 | ||
| AK126258.1 | 15 | 1957 | ||
| AK301100.1 | ||||
| AL512708.1 | 16 | 2005 | ||
| DQ786315.1 | 4 | 661 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833546 | Chr.17:76003077 - 76186972 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map