Assay Details
Target Gene Details
Entrez Gene ID: | 10351 |
Gene Name: | ATP binding cassette subfamily A member 8 |
Gene Aliases: |
- |
Location: |
Chr.17:68867285-68955426 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCA8 | NM_001288985.1 | 21 | 3048 | NP_001275914.1 |
| NM_001288986.1 | 20 | 2887 | NP_001275915.1 | |
| NM_007168.3 | 19 | 2767 | NP_009099.1 | |
| XM_005256938.2 | 20 | 2915 | XP_005256995.1 | |
| XM_005256940.4 | 19 | 2820 | XP_005256997.1 | |
| XM_011524191.2 | 20 | 2878 | XP_011522493.1 | |
| XM_011524192.2 | 20 | 2878 | XP_011522494.1 | |
| AB020629.1 | 19 | 2726 | ||
| AK295876.1 | 20 | 2860 | ||
| AK304286.1 | 19 | 2719 | ||
| AK304360.1 | ||||
| BC037309.1 | 21 | 2815 | AAH37309.1 | |
| BC130280.1 | 21 | 2897 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066412 | Chr.17:68901399 - 68957926 on Build GRCh38 | Loss |
|
| nsv1061151 | Chr.17:68885476 - 68977776 on Build GRCh38 | Gain |
|
| nsv575928 | Chr.17:68898720 - 68917795 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map