Assay Details
Target Gene Details
Entrez Gene ID: | 283459 |
Gene Name: | glutamyl-tRNA amidotransferase subunit C |
Gene Aliases: |
15E1.2 |
Location: |
Chr.12:120446438-120463753 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GATC | NM_176818.2 | 2 | 231 | NP_789788.1 |
| NR_033684.1 | 2 | 231 | ||
| AK094319.1 | 2 | 225 | ||
| BC107145.1 | 2 | 188 | AAI07146.1 | |
| BC107146.1 | 2 | 181 | AAI07147.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3630923 | Chr.12:120439963 - 120450931 on Build GRCh38 | Gain |
|
| nsv509482 | Chr.12:120435117 - 120495113 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map