Assay Details
Target Gene Details
Entrez Gene ID: | 51056 |
Gene Name: | leucine aminopeptidase 3 |
Gene Aliases: |
HEL-S-106, LAP, LAPEP, PEPS |
Location: |
Chr.4:17577304-17607967 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAP3 | NM_015907.2 | NP_056991.2 | ||
| AF061738.1 | AAD17527.1 | |||
| AK022055.1 | ||||
| AK225511.1 | ||||
| AK298613.1 | ||||
| AK298790.1 | ||||
| AK307723.1 | ||||
| BC006199.2 | AAH06199.3 | |||
| BC065564.1 | AAH65564.1 | |||
| BI827557.1 | ||||
| BM800626.1 | ||||
| CR457128.1 | CAG33409.1 | |||
| FJ224308.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv593781 | Chr.4:17030351 - 17591490 on Build GRCh38 | Gain |
|
| nsv289804 | Chr.4:17573750 - 17580165 on Build GRCh38 | Deletion |
|
| esv2759228 | Chr.4:17533621 - 17586697 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map