Assay Details
Target Gene Details
Entrez Gene ID: | 5281 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class F |
Gene Aliases: |
- |
Location: |
Chr.2:46581274-46617112 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGF | NM_002643.3 | NP_002634.1 | ||
| NM_173074.2 | NP_775097.1 | |||
| XM_011532908.2 | XP_011531210.1 | |||
| AK291108.1 | ||||
| BC021725.1 | AAH21725.1 | |||
| BC029408.1 | AAH29408.1 | |||
| CR457000.1 | CAG33281.1 | |||
| D13435.1 | BAA02697.1 |
Target Gene Details
Entrez Gene ID: | 23433 |
Gene Name: | ras homolog family member Q |
Gene Aliases: |
ARHQ, HEL-S-42, RASL7A, TC10, TC10A |
Location: |
Chr.2:46542409-46584688 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOQ | NM_012249.3 | 5 | 3490 | NP_036381.2 |
| EU794657.1 | 5 | 3490 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759046 | Chr.2:46305673 - 46647012 on Build GRCh38 | Loss |
|
| dgv149e55 | Chr.2:46378520 - 46591853 on Build GRCh38 | Loss |
|
| nsv1009735 | Chr.2:46497384 - 46686013 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map