Assay Details
Target Gene Details
Entrez Gene ID: | 23059 |
Gene Name: | clusterin associated protein 1 |
Gene Aliases: |
CFAP22, FAP22, IFT38 |
Location: |
Chr.16:3495562-3539048 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLUAP1 | NM_015041.2 | 2 | 211 | NP_055856.1 |
| XM_006720867.3 | XP_006720930.1 | |||
| XM_017023068.1 | 2 | 226 | XP_016878557.1 | |
| XM_017023069.1 | 2 | 226 | XP_016878558.1 | |
| XM_017023070.1 | 2 | 211 | XP_016878559.1 | |
| XM_017023071.1 | 2 | 1200 | XP_016878560.1 | |
| XM_017023072.1 | 2 | 1200 | XP_016878561.1 | |
| XM_017023073.1 | XP_016878562.1 | |||
| AB014543.1 | 2 | 138 | BAA31618.1 | |
| AB089691.1 | 2 | 193 | BAD44779.1 | |
| BC017070.2 | 2 | 139 | AAH17070.3 | |
| DA502615.1 | 2 | 214 |
More Information
Set Membership: |
|

Genomic Map