Assay Details
Target Gene Details
Entrez Gene ID: | 2153 |
Gene Name: | coagulation factor V |
Gene Aliases: |
FVL, PCCF, RPRGL1, THPH2 |
Location: |
Chr.1:169511954-169586632 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F5 | NM_000130.4 | NP_000121.2 | ||
| XM_017000660.1 | XP_016856149.1 | |||
| AK226079.1 | ||||
| DB182033.1 | ||||
| M14335.1 | AAB59532.1 | |||
| M16967.1 | AAA52424.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv22143 | Chr.1:169090390 - 169652030 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map