Assay Details
Target Gene Details
Entrez Gene ID: | 148 |
Gene Name: | adrenoceptor alpha 1A |
Gene Aliases: |
ADRA1C, ADRA1L1, ALPHA1AAR |
Location: |
Chr.8:26738113-26870994 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADRA1A | NM_001322502.1 | 2 | 1609 | NP_001309431.1 |
| NM_001322503.1 | NP_001309432.1 | |||
| NM_033302.3 | 3 | 1995 | NP_150645.2 | |
| NM_033303.4 | NP_150646.3 | |||
| NR_136343.1 | ||||
| XM_006716292.3 | XP_006716355.1 | |||
| XM_006716293.3 | 4 | 2330 | XP_006716356.1 | |
| XM_017013094.1 | XP_016868583.1 | |||
| XM_017013095.1 | XP_016868584.1 | |||
| AY491775.1 | AAR84644.1 | |||
| AY491776.1 | AAR84645.1 | |||
| AY491777.1 | 3 | 1534 | AAR84646.1 | |
| AY491778.1 | 2 | 1359 | AAR84647.1 | |
| D32201.1 | 3 | 1745 | BAA06900.1 | |
| D32202.1 | BAA06901.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610864 | Chr.8:26754119 - 26778227 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map