Assay Details
Target Gene Details
Entrez Gene ID: | 3309 |
Gene Name: | heat shock protein family A (Hsp70) member 5 |
Gene Aliases: |
BIP, GRP78, HEL-S-89n, MIF2 |
Location: |
Chr.9:125234848-125241387 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPA5 | NM_005347.4 | 8 | 3238 | NP_005338.1 |
| BC020235.1 | 8 | 3181 | AAH20235.1 |
More Information
Set Membership: |
|

Genomic Map