Assay Details
Target Gene Details
Entrez Gene ID: | 9117 |
Gene Name: | SEC22 homolog C, vesicle trafficking protein |
Gene Aliases: |
SEC22L3 |
Location: |
Chr.3:42547967-42601080 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC22C | NM_001201572.1 | NP_001188501.1 | ||
| BX377531.2 | ||||
| DA434403.1 |
Target Gene Details
Entrez Gene ID: | 51188 |
Gene Name: | SS18 like 2 |
Gene Aliases: |
KIAA-iso |
Location: |
Chr.3:42581840-42594998 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SS18L2 | NM_016305.3 | NP_057389.1 | ||
| XM_017006548.1 | XP_016862037.1 | |||
| XM_017006549.1 | XP_016862038.1 | |||
| AF201950.1 | AAF17242.1 | |||
| AK312225.1 | ||||
| BC017804.1 | AAH17804.1 | |||
| BX114027.1 | ||||
| DR158587.1 | ||||
| EL954786.1 |
More Information
Set Membership: |
|

Genomic Map