Assay Details
Target Gene Details
Entrez Gene ID: | 9650 |
Gene Name: | mitochondrial fission regulator 1 |
Gene Aliases: |
CHPPR, FAM54A2 |
Location: |
Chr.8:65643873-65771252 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTFR1 | NM_001145838.1 | NP_001139310.1 | ||
| NM_001145839.1 | NP_001139311.1 | |||
| NM_014637.3 | NP_055452.3 | |||
| XM_006716484.2 | XP_006716547.2 | |||
| XM_011517626.1 | XP_011515928.1 | |||
| XM_011517627.2 | XP_011515929.1 | |||
| AA015794.1 | ||||
| AK298945.1 | ||||
| AK304716.1 | ||||
| AK316202.1 | ||||
| BC036116.1 | AAH36116.1 | |||
| BC043498.2 | AAH43498.1 | |||
| BX537854.1 | CAD97862.1 | |||
| BX647507.1 | ||||
| D13634.1 | BAA02798.1 |
More Information
Set Membership: |
|

Genomic Map