Assay Details
Target Gene Details
Entrez Gene ID: | 345651 |
Gene Name: | actin, beta like 2 |
Gene Aliases: |
ACT |
Location: |
Chr.5:57480016-57482809 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTBL2 | NM_001017992.3 | 1 | 642 | NP_001017992.1 |
| BC137470.1 | 1 | 560 | ||
| BC137473.1 | 1 | 560 | ||
| BX648504.1 | 1 | 643 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3605137 | Chr.5:57481633 - 57484422 on Build GRCh38 | Loss |
|
| esv3575843 | Chr.5:57383265 - 58030187 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map