Assay Details
Target Gene Details
Entrez Gene ID: | 220965 |
Gene Name: | family with sequence similarity 13 member C |
Gene Aliases: |
FAM13C1 |
Location: |
Chr.10:59245933-59363186 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM13C | NM_001001971.2 | 12 | 1796 | NP_001001971.1 |
NM_001143773.1 | 15 | 2261 | NP_001137245.1 | |
NM_001166698.1 | 15 | 2172 | NP_001160170.1 | |
NM_198215.3 | 14 | 2090 | NP_937858.2 | |
XM_005269618.4 | 14 | 2087 | XP_005269675.2 | |
XM_005269619.4 | 12 | 1793 | XP_005269676.2 | |
XM_006717702.3 | 16 | 2091 | XP_006717765.1 | |
XM_006717703.3 | 16 | 2273 | XP_006717766.1 | |
XM_011539490.2 | 15 | 2146 | XP_011537792.1 | |
XM_011539491.2 | 16 | 2192 | XP_011537793.1 | |
XM_017015885.1 | 15 | 2004 | XP_016871374.1 | |
XM_017015886.1 | 15 | 2258 | XP_016871375.1 | |
XM_017015887.1 | 15 | 2008 | XP_016871376.1 | |
XM_017015888.1 | 16 | 2270 | XP_016871377.1 | |
XM_017015889.1 | 16 | 2088 | XP_016871378.1 | |
XM_017015890.1 | 16 | 2327 | XP_016871379.1 | |
XM_017015891.1 | 13 | 1967 | XP_016871380.1 | |
XM_017015892.1 | 13 | 1964 | XP_016871381.1 | |
XM_017015893.1 | 15 | 2068 | XP_016871382.1 | |
AK289796.1 | 14 | 2016 | ||
AK316542.1 | 15 | 2168 | ||
BC036453.1 | 14 | 2048 | AAH36453.1 | |
BC064431.1 | 12 | 1796 | AAH64431.1 | |
BM714192.1 |
Target Gene Details
Entrez Gene ID: | 84457 |
Gene Name: | phytanoyl-CoA 2-hydroxylase interacting protein like |
Gene Aliases: |
- |
Location: |
Chr.10:59174811-59247774 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHYHIPL | NM_001143774.1 | 5 | 3002 | NP_001137246.1 |
NM_032439.3 | 5 | 3222 | NP_115815.2 | |
XM_011540275.2 | 6 | 3284 | XP_011538577.1 | |
XM_011540276.2 | 5 | 4115 | XP_011538578.1 | |
XM_017016782.1 | 6 | 3103 | XP_016872271.1 | |
XM_017016783.1 | 6 | 4206 | XP_016872272.1 | |
AB058699.1 | 3 | 2563 | BAB47425.1 | |
AW299583.1 | 1 | 362 | ||
BM676577.1 | 1 | 369 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831888 | Chr.10:59183895 - 59378493 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
