Assay Details
Target Gene Details
Entrez Gene ID: | 2981 |
Gene Name: | guanylate cyclase activator 2B |
Gene Aliases: |
GCAP-II, UGN |
Location: |
Chr.1:42153421-42155824 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCA2B | NM_007102.2 | NP_009033.1 | ||
| BC069301.1 | AAH69301.1 | |||
| BC093779.1 | AAH93779.1 | |||
| BC093781.1 | AAH93781.1 | |||
| U34279.1 | AAC50416.1 | |||
| Z50753.1 | CAA90629.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527531 | Chr.1:42151361 - 42169834 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map