Assay Details
Target Gene Details
Entrez Gene ID: | 5036 |
Gene Name: | proliferation-associated 2G4 |
Gene Aliases: |
EBP1, HG4-1, p38-2G4 |
Location: |
Chr.12:56104319-56113910 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PA2G4 | NM_006191.2 | NP_006182.2 | ||
| AK291796.1 | ||||
| BC001951.1 | AAH01951.1 | |||
| BC007561.1 | AAH07561.1 | |||
| BC019222.1 | ||||
| BC032111.2 | AAH32111.1 | |||
| BC069786.1 | AAH69786.1 | |||
| BC072007.1 | AAH72007.1 | |||
| U59435.1 | AAB91536.1 | |||
| U87954.1 | AAD00646.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv973075 | Chr.12:56106326 - 56113947 on Build GRCh38 | Duplication |
|
| nsv1051961 | Chr.12:55868436 - 56235643 on Build GRCh38 | Gain |
|
| nsv832427 | Chr.12:55998761 - 56118990 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map