Assay Details
Target Gene Details
Entrez Gene ID: | 9792 |
Gene Name: | SERTA domain containing 2 |
Gene Aliases: |
Sei-2, TRIP-Br2 |
Location: |
Chr.2:64631621-64751091 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERTAD2 | NM_014755.2 | 2 | 691 | NP_055570.1 |
| XM_005264669.2 | 2 | 1209 | XP_005264726.1 | |
| XM_011533204.1 | 2 | 428 | XP_011531506.1 | |
| XM_011533205.2 | 2 | 745 | XP_011531507.1 | |
| AK311117.1 | ||||
| BC074788.2 | 2 | 436 | AAH74788.1 | |
| BC074789.2 | 2 | 436 | AAH74789.1 | |
| BC101639.1 | 2 | 475 | AAI01640.1 | |
| BC101641.1 | 2 | 476 | AAI01642.1 | |
| D50917.1 | 2 | 691 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582127 | Chr.2:63618350 - 64923736 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map