Assay Details
Target Gene Details
Entrez Gene ID: | 51340 |
Gene Name: | crooked neck pre-mRNA splicing factor 1 |
Gene Aliases: |
CLF, CRN, Clf1, HCRN, MSTP021, SYF3 |
Location: |
Chr.20:20034361-20056046 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRNKL1 | NM_001278625.1 | 15 | 4256 | NP_001265554.1 |
| NM_001278626.1 | 15 | 4069 | NP_001265555.1 | |
| NM_001278627.1 | 15 | 3979 | NP_001265556.1 | |
| NM_001278628.1 | 14 | 3927 | NP_001265557.1 | |
| NM_016652.5 | 15 | 4292 | NP_057736.4 | |
| AA255987.1 | 1 | 127 | ||
| AL832435.1 | 14 | 3901 | CAH10656.1 |
More Information
Set Membership: |
|

Genomic Map