Assay Details
Target Gene Details
Entrez Gene ID: | 79866 |
Gene Name: | bora, aurora kinase A activator |
Gene Aliases: |
C13orf34 |
Location: |
Chr.13:72727739-72756198 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BORA | NM_001286746.1 | 2 | 396 | NP_001273675.1 |
| NM_001286747.1 | 2 | 298 | NP_001273676.1 | |
| NM_024808.3 | 2 | 298 | NP_079084.3 | |
| XM_006719868.3 | 2 | 308 | XP_006719931.2 | |
| AK291496.1 | 2 | 124 | ||
| AK298608.1 | 2 | 124 | ||
| AK300758.1 | 2 | 143 | ||
| BC025367.2 | 2 | 109 | AAH25367.2 | |
| BC056876.1 | 2 | 121 | AAH56876.2 | |
| BM923278.1 | 2 | 307 | ||
| BX648605.1 | 2 | 396 | ||
| EU834129.1 | 2 | 204 |
More Information
Set Membership: |
|

Genomic Map