Assay Details
Target Gene Details
Entrez Gene ID: | 148867 |
Gene Name: | solute carrier family 30 member 7 |
Gene Aliases: |
ZNT7, ZnT-7, ZnTL2 |
Location: |
Chr.1:100896076-100996913 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC30A7 | NM_001144884.1 | 8 | 944 | NP_001138356.1 |
| NM_133496.4 | 8 | 942 | NP_598003.2 | |
| XM_011540779.2 | 9 | 1041 | XP_011539081.1 | |
| XM_017000400.1 | 8 | 942 | XP_016855889.1 | |
| XM_017000401.1 | 8 | 942 | XP_016855890.1 | |
| AF233345.1 | 8 | 772 | AAL83716.1 | |
| AF529197.1 | 8 | 757 | AAO17324.1 | |
| AK313638.1 | 8 | 925 | ||
| AY094606.1 | 8 | 757 | AAM21969.1 | |
| BC064692.1 | 8 | 898 | AAH64692.1 | |
| BX537375.1 | 8 | 944 | CAD97617.1 | |
| CR936806.1 | 8 | 938 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522064 | Chr.1:100715786 - 101074310 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map